Assay Details
| Gene Symbol: ERP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195735.1 | NP_001182664.1 | 151 | 71 | ||
| GenBank mRNA | CO648391.1 | - | 155 | 71 | ||
| JU474098.1 | - | 215 | 71 | |||
| JV046023.1 | - | 234 | 71 | |||
| Gene Symbol: LOC720396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028830377.1 | XP_028686210.1 | 155 | 71 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ERP29
Genomic Map