Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801521.3 | XP_002801567.1 | 1287 | 71 | ||
| XM_015138048.2 | XP_014993534.1 | 1183 | 71 | |||
| XM_015138059.2 | XP_014993545.1 | 1062 | 71 | |||
| GenBank mRNA | JU473205.1 | - | 1199 | 71 | ||
| JV050297.1 | - | 569 | 71 | |||
| JV635424.1 | - | 1154 | 71 | |||
More Information
Set Membership: |
|

KIF2C
Genomic Map