Assay Details
| Gene Symbol: PIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265848.1 | NP_001252777.1 | 337 | 69 | ||
| GenBank mRNA | JU326868.1 | - | 103 | 69 | ||
| JU474115.1 | - | 370 | 69 | |||
| JV046040.1 | - | 337 | 69 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PIM2
Genomic Map