Assay Details
| Gene Symbol: RP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267788.1 | NP_001254717.1 | 918 | 131 | ||
| GenBank mRNA | JU321774.1 | - | 782 | 131 | ||
| JU476692.1 | - | 918 | 131 | |||
More Information
Set Membership: |
|

RP2
Genomic Map