Assay Details
| Gene Symbol: ST3GAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266341.1 | NP_001253270.1 | 1964 | 115 | ||
| XM_028840536.1 | XP_028696369.1 | 2065 | 115 | |||
| XM_028840537.1 | XP_028696370.1 | 1869 | 115 | |||
| GenBank mRNA | JU332128.1 | - | 1834 | 115 | ||
| JU470852.1 | - | 1973 | 115 | |||
| JV044250.1 | - | 1964 | 115 | |||
| JV567663.1 | - | 2021 | 115 | |||
More Information
Set Membership: |
|

ST3GAL2
Genomic Map