Assay Details
| Gene Symbol: MMP23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095296.4 | XP_001095296.2 | 958 | 110 | ||
| GenBank mRNA | JV044396.1 | - | 468 | 110 | ||
| JV663997.1 | - | 413 | 110 | |||
More Information
Set Membership: |
|

MMP23B
Genomic Map