Assay Details
| Gene Symbol: LRRC23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110883.4 | XP_001110883.1 | 1314 | 116 | ||
| XM_028829216.1 | XP_028685049.1 | 1324 | 116 | |||
| XM_028829218.1 | XP_028685051.1 | 1325 | 116 | |||
| XM_028829219.1 | XP_028685052.1 | 1204 | 116 | |||
| XM_028829220.1 | XP_028685053.1 | 1215 | 116 | |||
| GenBank mRNA | JU337624.1 | - | 346 | 116 | ||
| JV705024.1 | - | 254 | 116 | |||
More Information
Set Membership: |
|

LRRC23
Genomic Map