Assay Details
| Gene Symbol: SLC19A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257523.1 | NP_001244452.1 | 1421 | 74 | ||
| XM_028835438.1 | XP_028691271.1 | 1783 | 74 | |||
| XM_028835442.1 | XP_028691275.1 | 968 | 74 | |||
| GenBank mRNA | JU332537.1 | - | 1421 | 74 | ||
| JU474013.1 | - | 1401 | 74 | |||
| JV045940.1 | - | 1558 | 74 | |||
More Information
Set Membership: |
|

SLC19A2
Genomic Map