Assay Details
| Gene Symbol: TPPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266695.1 | NP_001253624.1 | 561 | 112 | ||
| XM_015139672.2 | XP_014995158.2 | 884 | 112 | |||
| XM_028849237.1 | XP_028705070.1 | 489 | 112 | |||
| XM_028849238.1 | XP_028705071.1 | 585 | 112 | |||
| XM_028849239.1 | XP_028705072.1 | 544 | 112 | |||
| GenBank mRNA | JU332683.1 | - | 561 | 112 | ||
| JV566132.1 | - | 580 | 112 | |||
More Information
Set Membership: |
|

TPPP
Genomic Map