Assay Details
| Gene Symbol: ADAMTS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845389.1 | XP_028701222.1 | 2337 | 74 | ||
| XM_028845390.1 | XP_028701223.1 | 2287 | 74 | |||
More Information
Set Membership: |
|

ADAMTS5
Genomic Map