Assay Details
| Gene Symbol: RPP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266712.1 | NP_001253641.1 | 540 | 108 | ||
| XM_015130593.2 | XP_014986079.1 | 834 | 108 | |||
| XM_015130594.2 | XP_014986080.1 | 605 | 108 | |||
| XM_015130595.2 | XP_014986081.1 | 500 | 108 | |||
| XM_015130596.2 | XP_014986082.1 | 430 | 108 | |||
| XM_015130597.2 | XP_014986083.1 | 837 | 108 | |||
| XM_028843253.1 | XP_028699086.1 | 647 | 108 | |||
| XM_028843254.1 | XP_028699087.1 | 624 | 108 | |||
| GenBank mRNA | JU332688.1 | - | 497 | 108 | ||
| JU472057.1 | - | 370 | 108 | |||
| JU472058.1 | - | 714 | 108 | |||
| JV044948.1 | - | 540 | 108 | |||
| JV044949.1 | - | 507 | 108 | |||
| JV044950.1 | - | 540 | 108 | |||
More Information
Set Membership: |
|

RPP14
Genomic Map