Assay Details
| Gene Symbol: LOC114677617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847619.1 | XP_028703452.1 | 615 | 145 | ||
| GenBank mRNA | JV570193.1 | - | 322 | 145 | ||
| Gene Symbol: STK38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260590.1 | NP_001247519.1 | 1527 | 145 | ||
| GenBank mRNA | JU327690.1 | - | 303 | 145 | ||
| JU474200.1 | - | 1527 | 145 | |||
| JV455740.1 | - | 301 | 145 | |||
More Information
Set Membership: |
|

LOC114677617
Genomic Map