Assay Details
| Gene Symbol: TUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261666.1 | NP_001248595.1 | 239 | 152 | ||
| GenBank mRNA | JU332785.1 | - | 258 | 152 | ||
| JU474202.1 | - | 239 | 152 | |||
| JV532346.1 | - | 258 | 152 | |||
More Information
Set Membership: |
|

TUSC2
Genomic Map