Assay Details
| Gene Symbol: NUP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266179.1 | NP_001253108.1 | 443 | 84 | ||
| GenBank mRNA | JU332685.1 | - | 420 | 84 | ||
| JU474129.1 | - | 443 | 84 | |||
More Information
Set Membership: |
|

NUP42
Genomic Map