Assay Details
| Gene Symbol: GPR161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850124.1 | XP_028705957.1 | 1220 | 126 | ||
| XM_028850125.1 | XP_028705958.1 | 1397 | 126 | |||
| XM_028850126.1 | XP_028705959.1 | 1377 | 126 | |||
| XM_028850127.1 | XP_028705960.1 | 1781 | 126 | |||
| XM_028850128.1 | XP_028705961.1 | 1127 | 126 | |||
| GenBank mRNA | JV056427.1 | - | 528 | 126 | ||
More Information
Set Membership: |
|

GPR161
Genomic Map