Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266704.1 | NP_001253633.1 | 749 | 91 | ||
| XM_028844368.1 | XP_028700201.1 | 374 | 91 | |||
| XM_028844369.1 | XP_028700202.1 | 358 | 91 | |||
| GenBank mRNA | JU333735.1 | - | 749 | 91 | ||
| JV602203.1 | - | 750 | 91 | |||
More Information
Set Membership: |
|

DDAH1
Genomic Map