Assay Details
| Gene Symbol: GTF3C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260973.1 | NP_001247902.1 | 2280 | 140 | ||
| GenBank mRNA | JU327603.1 | - | 2054 | 140 | ||
| JU477867.1 | - | 2280 | 140 | |||
| JV727087.1 | - | 2095 | 140 | |||
More Information
Set Membership: |
|

GTF3C4
Genomic Map