Assay Details
| Gene Symbol: MLYCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841137.1 | XP_028696970.1 | 599 | 87 | ||
| GenBank mRNA | JU333686.1 | - | 549 | 87 | ||
| JV611149.1 | - | 545 | 87 | |||
More Information
Set Membership: |
|

MLYCD
Genomic Map