Assay Details
| Gene Symbol: MMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194810.2 | NP_001181739.1 | 1789 | 170 | ||
| XM_001103680.4 | XP_001103680.1 | 1952 | 170 | |||
| GenBank mRNA | JU319842.1 | - | 1933 | 170 | ||
| JV044241.1 | - | 1904 | 170 | |||
| JV668483.1 | - | 1936 | 170 | |||
More Information
Set Membership: |
|

MMD
Genomic Map