Assay Details
| Gene Symbol: BAMBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146693.2 | XP_015002179.2 | 638 | 77 | ||
| GenBank mRNA | JU333848.1 | - | 215 | 77 | ||
| JV637452.1 | - | 322 | 77 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAMBI| Gene Symbol: BAMBI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146693.2 | XP_015002179.2 | 638 | 77 | ||
| GenBank mRNA | JU333848.1 | - | 215 | 77 | ||
| JV637452.1 | - | 322 | 77 | |||