Assay Details
| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837967.1 | XP_028693800.1 | 2804 | 98 | ||
| XM_028837968.1 | XP_028693801.1 | 2817 | 98 | |||
More Information
Set Membership: |
|

SERPINB13
Genomic Map