Assay Details
| Gene Symbol: RSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093095.4 | XP_001093095.4 | 833 | 97 | ||
| XM_028826023.1 | XP_028681856.1 | 952 | 97 | |||
| GenBank mRNA | JU320433.1 | - | 851 | 97 | ||
| JU320434.1 | - | 844 | 97 | |||
| JU471658.1 | - | 861 | 97 | |||
| JU471659.1 | - | 838 | 97 | |||
| JV043345.1 | - | 862 | 97 | |||
| JV043346.1 | - | 857 | 97 | |||
| JV668099.1 | - | 772 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RSU1
Genomic Map