Assay Details
| Gene Symbol: LOC719351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828559.1 | XP_028684392.1 | 1329 | 99 | ||
| GenBank mRNA | JU333764.1 | - | 2311 | 99 | ||
| JU475010.1 | - | 2357 | 99 | |||
| JV046633.1 | - | 2358 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC719351
Genomic Map