Assay Details
| Gene Symbol: LOC719351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828559.1 | XP_028684392.1 | 1798 | 124 | ||
| GenBank mRNA | JU333764.1 | - | 2780 | 124 | ||
| JU475010.1 | - | 2826 | 124 | |||
| JV046633.1 | - | 2827 | 124 | |||
More Information
Set Membership: |
|

LOC719351
Genomic Map