Assay Details
| Gene Symbol: WBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258099.2 | NP_001245028.1 | 709 | 107 | ||
| XM_015120332.2 | XP_014975818.1 | 1040 | 107 | |||
| GenBank mRNA | JU326603.1 | - | 525 | 107 | ||
| JU474974.1 | - | 709 | 107 | |||
| JV043584.1 | - | 718 | 107 | |||
| JV668423.1 | - | 773 | 107 | |||
More Information
Set Membership: |
|

WBP2
Genomic Map