Assay Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124228.2 | XP_014979714.2 | 547 | 124 | ||
| XM_015124229.2 | XP_014979715.2 | 307 | 124 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015124229.2 transcripts.
Set Membership: |
|

ZNF180
Genomic Map