Assay Details
| Gene Symbol: CCDC106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193843.3 | NP_001180772.2 | 491 | 112 | ||
| XM_001089505.3 | XP_001089505.2 | 865 | 112 | |||
| XM_028838394.1 | XP_028694227.1 | 355 | 112 | |||
| GenBank mRNA | JU334209.1 | - | 271 | 112 | ||
| JV649529.1 | - | 578 | 112 | |||
More Information
Set Membership: |
|

CCDC106
Genomic Map