Assay Details
| Gene Symbol: TFPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194720.3 | NP_001181649.3 | 408 | 73 | ||
| GenBank mRNA | JU321409.1 | - | 412 | 73 | ||
| JU473608.1 | - | 348 | 73 | |||
| JV045754.1 | - | 285 | 73 | |||
| JV566358.1 | - | 419 | 73 | |||
| JV669731.1 | - | 328 | 73 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194720.3 transcripts.
Set Membership: |
|

TFPT
Genomic Map