Assay Details
| Gene Symbol: ZNF225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801274.3 | XP_002801320.2 | 208 | 138 | ||
| GenBank mRNA | JU323829.1 | - | 152 | 138 | ||
| JU493113.1 | - | 160 | 138 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002801274.3 transcripts.
Set Membership: |
|

ZNF225
Genomic Map