Assay Details
| Gene Symbol: SLC25A24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258085.1 | NP_001245014.1 | 866 | 89 | ||
| XM_015146189.2 | XP_015001675.2 | 795 | 89 | |||
| GenBank mRNA | JU471891.1 | - | 866 | 89 | ||
More Information
Set Membership: |
|

SLC25A24
Genomic Map