Assay Details
| Gene Symbol: PREB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266941.1 | NP_001253870.1 | 868 | 76 | ||
| XM_028831459.1 | XP_028687292.1 | 887 | 76 | |||
| GenBank mRNA | JU332408.1 | - | 829 | 76 | ||
| JU473874.1 | - | 853 | 76 | |||
| JV045839.1 | - | 868 | 76 | |||
| JV670009.1 | - | 875 | 76 | |||
More Information
Set Membership: |
|

PREB
Genomic Map