Assay Details
| Gene Symbol: ADGRE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038662.1 | NP_001033751.1 | 2192 | 123 | ||
| XM_028838330.1 | XP_028694163.1 | 2346 | 123 | |||
| XM_028838331.1 | XP_028694164.1 | 2346 | 123 | |||
| XM_028838333.1 | XP_028694166.1 | 2202 | 123 | |||
| GenBank mRNA | DQ227271.1 | - | 2192 | 123 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADGRE2
Genomic Map