Assay Details
| Gene Symbol: DSPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098430.4 | XP_001098430.2 | 172 | 147 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSPP| Gene Symbol: DSPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098430.4 | XP_001098430.2 | 172 | 147 | ||
Set Membership: |
|