Assay Details
| Gene Symbol: C14H11orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193864.2 | NP_001180793.1 | 485 | 150 | ||
| GenBank mRNA | JU332649.1 | - | 355 | 150 | ||
| JU472400.1 | - | 448 | 150 | |||
| JV637307.1 | - | 437 | 150 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C14H11orf58
Genomic Map