Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261063.2 | NP_001247992.1 | 494 | 113 | ||
| XM_015125580.2 | XP_014981066.1 | 412 | 113 | |||
| XM_015125581.2 | XP_014981067.1 | 382 | 113 | |||
| XM_028840534.1 | XP_028696367.1 | 387 | 113 | |||
| GenBank mRNA | JU332254.1 | - | 466 | 113 | ||
| JU471565.1 | - | 630 | 113 | |||
| JV044493.1 | - | 496 | 113 | |||
| JV638011.1 | - | 571 | 113 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CARHSP1
Genomic Map