Assay Details
| Gene Symbol: ZBTB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097102.4 | XP_001097102.1 | 1250 | 88 | ||
| XM_015123688.2 | XP_014979174.1 | 1252 | 88 | |||
| XM_015123693.2 | XP_014979179.1 | 307 | 88 | |||
| XM_028839238.1 | XP_028695071.1 | 313 | 88 | |||
More Information
Set Membership: |
|

ZBTB32
Genomic Map