Assay Details
| Gene Symbol: LSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145176.2 | XP_015000662.1 | 445 | 71 | ||
| GenBank mRNA | JU334018.1 | - | 216 | 71 | ||
| JU472804.1 | - | 220 | 71 | |||
| JV045444.1 | - | 166 | 71 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LSM1
Genomic Map