Assay Details
| Gene Symbol: DHDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124512.2 | XP_014979998.1 | 418 | 90 | ||
| GenBank mRNA | JU323376.1 | - | 402 | 90 | ||
| JU478319.1 | - | 392 | 90 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DHDH
Genomic Map