Assay Details
| Gene Symbol: TP53TG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194416.2 | NP_001181345.1 | 410 | 141 | ||
| XM_015148829.2 | XP_015004315.1 | 316 | 141 | |||
| XM_028827330.1 | XP_028683163.1 | 425 | 141 | |||
More Information
Set Membership: |
|

TP53TG5
Genomic Map