Assay Details
| Gene Symbol: ZNF232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118380.2 | XP_014973866.2 | 1138 | 105 | ||
| GenBank mRNA | JU323292.1 | - | 651 | 105 | ||
| JU473404.1 | - | 651 | 105 | |||
More Information
Set Membership: |
|

ZNF232
Genomic Map