Assay Details
| Gene Symbol: MAD2L1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265944.1 | NP_001252873.1 | 337 | 141 | ||
| GenBank mRNA | JU323314.1 | - | 357 | 141 | ||
| JU472623.1 | - | 337 | 141 | |||
More Information
Set Membership: |
|

MAD2L1BP
Genomic Map