Assay Details
| Gene Symbol: NOS1AP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164764.3 | NP_001158236.1 | 706 | 98 | ||
| XM_028846397.1 | XP_028702230.1 | 1103 | 98 | |||
| XM_028846398.1 | XP_028702231.1 | 980 | 98 | |||
| XM_028846399.1 | XP_028702232.1 | 952 | 98 | |||
| XM_028846400.1 | XP_028702233.1 | 6046 | 98 | |||
| GenBank mRNA | JU320195.1 | - | 706 | 98 | ||
| JU328629.1 | - | 356 | 98 | |||
| JV599433.1 | - | 578 | 98 | |||
More Information
Set Membership: |
|

NOS1AP
Genomic Map