Assay Details
| Gene Symbol: ENPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266236.1 | NP_001253165.1 | 1045 | 97 | ||
| XM_015136457.2 | XP_014991943.1 | 1307 | 97 | |||
| GenBank mRNA | JU322749.1 | - | 1223 | 97 | ||
| JU471796.1 | - | 1045 | 97 | |||
More Information
Set Membership: |
|

ENPP4
Genomic Map