Assay Details
| Gene Symbol: SARM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265713.1 | NP_001252642.1 | 1464 | 89 | ||
| XM_028835706.1 | XP_028691539.1 | 1391 | 89 | |||
| GenBank mRNA | JU327461.1 | - | 390 | 89 | ||
| JU327462.1 | - | 390 | 89 | |||
| JU327463.1 | - | 390 | 89 | |||
| JV600183.1 | - | 1464 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SARM1
Genomic Map