Assay Details
| Gene Symbol: NEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152077.2 | XP_015007563.1 | 166 | 79 | ||
| XM_015152078.1 | XP_015007564.1 | 166 | 79 | |||
| XM_015152079.2 | XP_015007565.1 | 166 | 79 | |||
| XM_015152080.2 | XP_015007566.1 | 166 | 79 | |||
More Information
Set Membership: |
|

NEMP1
Genomic Map