Assay Details
| Gene Symbol: SYNM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108469.4 | XP_001108469.3 | 1090 | 106 | ||
| XM_015143168.2 | XP_014998654.2 | 1091 | 106 | |||
| GenBank mRNA | JU333649.1 | - | 1054 | 106 | ||
| JU333650.1 | - | 1054 | 106 | |||
More Information
Set Membership: |
|

SYNM
Genomic Map