Assay Details
| Gene Symbol: ADNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266818.1 | NP_001253747.1 | 920 | 69 | ||
| GenBank mRNA | JU333673.1 | - | 203 | 69 | ||
| JU474928.1 | - | 203 | 69 | |||
| JV046585.1 | - | 920 | 69 | |||
| JV046586.1 | - | 920 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266818.1 transcripts.
Set Membership: |
|

ADNP
Genomic Map