Assay Details
| Gene Symbol: GORASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261707.2 | NP_001248636.1 | 947 | 64 | ||
| GenBank mRNA | JU333898.1 | - | 934 | 64 | ||
| JU475082.1 | - | 934 | 64 | |||
| JV044418.1 | - | 949 | 64 | |||
More Information
Set Membership: |
|

GORASP2
Genomic Map