Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266433.1 | NP_001253362.1 | 345 | 59 | ||
| GenBank mRNA | JU334733.1 | - | 388 | 59 | ||
| JU475784.1 | - | 345 | 59 | |||
| JV470133.1 | - | 369 | 59 | |||
More Information
Set Membership: |
|

CALY
Genomic Map