Assay Details
| Gene Symbol: LAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260698.1 | NP_001247627.1 | 1265 | 106 | ||
| GenBank mRNA | JU321430.1 | - | 1265 | 106 | ||
| JV639285.1 | - | 1328 | 106 | |||
More Information
Set Membership: |
|

LAP3
Genomic Map